missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A4 Antibody (SAIC-14C-10F12) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20093JF585
This item is not returnable.
View return policy
Description
Annexin A4 Monoclonal antibody specifically detects Annexin A4 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Mass Spectrometry,Immunocytochemistry/ Immunofluorescence
Specifications
| Annexin A4 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| annexin A4, Annexin IV, annexin IV (placental anticoagulant protein II), annexin-4, ANX4Carbohydrate-binding protein p33/p41, Chromobindin-4, chromobindin-4,35-beta calcimedin, DKFZp686H02120, Endonexin I, Lipocortin IV, MGC75105, P32.5, PAP-II, PIG28, Placental anticoagulant protein II, PP4-X, proliferation-inducing gene 28, proliferation-inducing protein 28, Protein II, ZAP36 | |
| Peptide 'DEGNYLDDALVR' derived from Annexin A4 conjugated to KLH. | |
| 0.1 mL | |
| Cancer | |
| 307 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, ELISA, Western Blot, Mass Spectrometry, Immunocytochemistry/Immunofluorescence | |
| SAIC-14C-10F12 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction