Learn More
Annexin A6 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98751APC
Product Code. 18634555
  • € 407.00 / 0.10mL
Description
Annexin A6 Polyclonal antibody specifically detects Annexin A6 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)