missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A6 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99419IR
This item is not returnable.
View return policy
Description
Annexin A6 Polyclonal antibody specifically detects Annexin A6 in Mouse samples. It is validated for Western Blot
Specifications
| Annexin A6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| 67 kDa calelectrin, annexin A6, Annexin VI, annexin VI (p68), annexin-6, ANX6, calcium-binding protein p68, calelectrin, calphobindin II, calphobindin-II, CBP68, Chromobindin-20, CPB-II, Lipocortin VI, p68, p70, Protein III | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Annexin A6 | |
| 0.1 mL | |
| Endosome Markers | |
| 309 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction