missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A7 Antibody (OTI1C3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70181
This item is not returnable.
View return policy
Description
Annexin A7 Monoclonal specifically detects Annexin A7 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Annexin A7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| annexin A7, ANX7, ANXA7, SNX, SYNEXIN | |
| Mouse | |
| 52.6 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI1C3 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| ANXA7 | |
| Human recombinant protein fragment corresponding to amino acids 105-411 of human ANXA7(NP_004025) produced in E.coli. | |
| Affinity Purified | |
| Cancer | |
| 310 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction