missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A7 Antibody (OTI1C3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70181G
This item is not returnable.
View return policy
Description
Annexin A7 Monoclonal specifically detects Annexin A7 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Annexin A7 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANXA7 | |
| Recombinant protein fragment corresponding to amino acids 105-411 of human Annexin A7 (NP_004025) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1C3 | |
| Western Blot | |
| annexin A7, ANX7, ANXA7, SNX, SYNEXIN | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 310.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction