missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A7 Antibody (S07-1D2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19944-50ul
This item is not returnable.
View return policy
Description
Annexin A7 Monoclonal antibody specifically detects Annexin A7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Annexin A7 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| annexin A7, ANX7, ANXA7, SNX, SYNEXIN | |
| Recombinant protein of human Annexin A7 (Uniprot # P20073) | |
| 50 μg | |
| Cancer | |
| 310 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S07-1D2 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction