missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A8/ANXA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97051-100ul
This item is not returnable.
View return policy
Description
Annexin A8/ANXA8 Polyclonal antibody specifically detects Annexin A8/ANXA8 in Human samples. It is validated for ELISA
Specifications
| Annexin A8/ANXA8 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| annexin A8, Annexin VIII, Annexin-8, ANX8, FLJ53095, VAC-beta, Vascular anticoagulant-beta | |
| Produced in rabbits immunized with purified, recombinant Human Annexin A8/ANXA8 (Accession#: AAH73755.1; Met1-Pro327) | |
| 100 μg | |
| Signal Transduction | |
| 653145 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction