missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Annexin V Antibody (03) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30742AF700
This item is not returnable.
View return policy
Description
Annexin V Monoclonal antibody specifically detects Annexin V in Human samples. It is validated for ELISA,Western Blot
Specifications
| Annexin V | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
| Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Annexin V (Uniprot#: P08758; Met1-Asp320) | |
| 0.1 mL | |
| Apoptosis, Apoptosis Detections | |
| 308 | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Western Blot | |
| 03 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction