Learn More
Novus Biologicals™ Annexin V Antibody (03) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30742MFV610
Product Code. 30042848
  • ONLINE EXCLUSIVE PRICE
    € 541.00
    € 511.35 / 0.10mL
    Save € 29.65
    5% Off
Description
Annexin V Monoclonal antibody specifically detects Annexin V in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Annexin V
Monoclonal
mFluor Violet 610 SE
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha
Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Annexin V (Uniprot#: P08758; Met1-Asp320)
0.1 mL
Apoptosis, Apoptosis Detections
308
Store at 4°C in the dark.
IgG2a
ELISA, Western Blot
03
50 mM Sodium Borate
Mouse
Protein A purified