missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin V Antibody (3B9F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 217.00 - € 557.00
Specifications
| Antigen | Annexin V |
|---|---|
| Clone | 3B9F3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261110
|
Novus Biologicals
NBP2-52413 |
0.1 mg |
€ 557.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663327
|
Novus Biologicals
NBP2-52413-0.025mg |
0.025 mg |
€ 217.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Annexin V Monoclonal specifically detects Annexin V in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Annexin V | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Apoptosis Detections | |
| PBS with 0.05% Sodium Azide | |
| 308 | |
| Purified recombinant fragment of human ANXA5 (AA: 160-320) expressed in E. Coli. | |
| Primary | |
| 36 kDa |
| 3B9F3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
| ANXA5 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title