missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANO1 Monoclonal Antibody (OTI5H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803579S
This item is not returnable.
View return policy
Description
ANO1 Monoclonal antibody specifically detects ANO1 in Human samples. It is validated for Western BlotSpecifications
| ANO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5XXA6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55107 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5H2 | |
| Unconjugated | |
| ANO1 | |
| ANO1; anoctamin 1; anoctamin 1, calcium activated chloride channel; anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; discovered on gastrointestinal stromal tumors protein 1; DOG1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; ORAOV2; TAOS2; Tmem16a; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); tumor-amplified and overexpressed sequence 2 | |
| Human recombinant protein fragment corresponding to amino acids 1-333 of human ANO1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction