Learn More
ANO1 Mouse anti-Human, Clone: OTI5H7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803578
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.Specifications
| ANO1 | |
| Monoclonal | |
| Unconjugated | |
| ANO1 | |
| DOG1, ORAOV2, TAOS2, TMEM16A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55107 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5H7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q5XXA6 | |
| ANO1 | |
| Human recombit protein fragment corresponding to amino acids 1-333 of human ANO1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.