missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANO2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-81703AF750
This item is not returnable.
View return policy
Description
ANO2 Polyclonal antibody specifically detects ANO2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ANO2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| anoctamin 2, anoctamin-2, DKFZp434P102, TMEM16B, transmembrane protein 16B (eight membrane-spanning domains) | |
| ANO2 antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human ANO2. The immunogen is located within the first 50 amino acids of ANO2. Amino Acid Squence: PRRLSPQAGSRGGQGPKHG | |
| 0.1 mL | |
| Neuroscience | |
| 57101 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction