missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANO6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535240
This item is not returnable.
View return policy
Description
ANO6 Polyclonal Antibody for Western Blot
May act as a calcium-activated chloride channel.
Specifications
| ANO6 | |
| Polyclonal | |
| Unconjugated | |
| ANO6 | |
| 2900059G15Rik; AA407480; Ano6; anoctamin 6; anoctamin-6; AW554778; BDPLT7; F730003B03Rik; LOW QUALITY PROTEIN: anoctamin-6; SCAN channel; SCTS; small-conductance calcium-activated nonselective cation channel; Tmem16f; Transmembrane protein 16F | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 196527 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q4KMQ2 | |
| ANO6 | |
| KLH conjugated synthetic peptide between 68-96 amino acids from the N-terminal region of human ANO6. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction