Learn More
Applied Biosystems™ Anode Buffer Container Dx
Product Code. 16315322 Shop All Applied Biosystems Products
Click to view available options
Quantity:
4 Ea.
Unit Size:
Pack of 4
Product Code. 16315322
Brand: Applied Biosystems™ 4393925
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Each package contains 4 buffer containers
The Anode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers.
The Anode Buffer Container Dx is designed for use with Applied Biosystems Dx genetic analyzers.
The Anode Buffer Container Dx contains 1X running buffer to support all electrophoresis applications on Applied Biosystems Dx genetic analyzers. It is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the Anode Buffer Container Dx is heat-sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.
This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.
This is an anode buffer container designed for use with the 3500 Dx/3500xL Dx Genetic Analyzer. The 3500 Dx series conditioning reagent, anode buffer, cathode buffer and septa cathode buffer are required for instrument operation.
Order Info
4 Pack
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Each package contains 4 buffer containers.
Store at 2 - 8°C.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
For Use With (Application) Sequencing
Product Type Anode Buffer Container
Quantity 4 Ea.
Shipping Condition Wet Ice
For In Vitro Diagnostic Use Only