Learn More
Emma Safety Footwear Anouk Safety Shoes
An attractive, feminine shoe that's supremely comfortable regardless of how hard the floor is. Unique SmartTecAir™ technology removes moisture and perspiration instantly. Anatomically designed and sculpted to the foot, the low profile shoe offers ESD certification and anti-fatigue properties.
Brand: Emma Safety Footwear MM94651636
Additional Details : Weight : 0.01000kg
Description
- S3-Safety shoe with sole plate for wet work
- Microfiber construction
- GripForce™ Easy Twist
- ESD Certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
36 |
Laces | |
Anouk | |
Microfiber | |
Steel |