missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANP32A Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-25097FR
This item is not returnable.
View return policy
Description
ANP32A Polyclonal antibody specifically detects ANP32A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
Specifications
| ANP32A | |
| Polyclonal | |
| Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
| acidic (leucine-rich) nuclear phosphoprotein 32 family, member A, acidic leucine-rich nuclear phosphoprotein 32 family member A, Acidic nuclear phosphoprotein pp32, cerebellar leucine rich acidic nuclear protein, I1PP2A, inhibitor-1 of protein phosphatase-2A, LANPPutative HLA-DR-associated protein I, Leucine-rich acidic nuclear protein, MAPMC15orf1, Mapmodulin, PHAP1MGC150373, PHAPIMGC119787, Potent heat-stable protein phosphatase 2A inhibitor I1PP2A, PP32, putative human HLA class II associated protein I | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human ANP32A. The immunogen is located within the last 50 amino acids of ANP32A. | |
| 0.1 mL | |
| Apoptosis | |
| 8125 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction