missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANP32A Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ANP32A |
|---|---|
| Clone | 2D8C5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235667
|
Proteintech
67687-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234615
|
Proteintech
67687-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ANP32A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ANP32A. This antibody recognizes human, mouse, rat antigen. The ANP32A antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| ANP32A | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 8125 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2D8C5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| C15orf1; I1PP2A; LANP; MAPM; MGC119787; MGC150373; PHAP1; PHAPI; PP32 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title