missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANP32B Rabbit anti-Human, Mouse, Rat, Clone: 4A8E5, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 190.00 - € 496.00
Specifications
| Antigen | ANP32B |
|---|---|
| Clone | 4A8E5 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359586
|
Novus Biologicals
NBP3-16199-20UL |
20 μg |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18378406
|
Novus Biologicals
NBP3-16199-100UL |
100 μg |
€ 496.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANP32B Monoclonal antibody specifically detects ANP32B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| ANP32B | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| acidic (leucine-rich) nuclear phosphoprotein 32 family, member B, acidic leucine-rich nuclear phosphoprotein 32 family member B, Acidic protein rich in leucines, APRILSilver-stainable protein SSP29, Pal31, PHAPI2SSP29, Putative HLA-DR-associated protein I-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ANP32B (Q92688). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4A8E5 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10541 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title