missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANR44 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15774261 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15774261 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15774261 Supplier Invitrogen™ Supplier No. PA556352

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TALHYAAASD MDRNKTILGN AHDNSEELER ARELKEKEAT LCLEFLLQND ANPSIR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

Putative regulatory subunit of protein phospatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANR44
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD44
Gene Accession No. Q8N8A2
Gene Alias ANKRD44; ankyrin repeat domain 44; ankyrin repeat domain-containing protein 44; PP6-ARS-B; protein phosphatase 6 ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory subunit ARS-B
Gene Symbols ANKRD44
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANR44. Recombinant protein control fragment (Product #RP-95964).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.