Learn More
Invitrogen™ ANR44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597101
Description
Immunogen sequence: LHMTAVHGRF TRSQTLIQNG GEIDCVDKDG NTPLHVAARY GHELLINTLI TSGADTAKCG IHSMFPLHLA ALNAHSDCCR KLLSSGFEID TPDKFGRTCL HAAAAGGNVE CIKLLQSSGA DFHKKDKCGR TPLHYAAANC HFHCIETLVT TGANVNETDD WGRTALHYAA ASDMDRNKTI LGNAHDNSEE LERARELKEK EATLCLEFLL QNDANPSIRD KEGYNSIHYA AAYGHRQCLE L; Positive Samples: H460, Jurkat.
Putative regulatory subunit of protein phospatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates.
Specifications
| ANR44 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD44 | |
| ANKRD44; ankyrin repeat domain 44; ankyrin repeat domain-containing protein 44; PP6-ARS-B; protein phosphatase 6 ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory subunit ARS-B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 91526 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.84 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N8A2 | |
| ANKRD44 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-550 of human ANKRD44 (NP_0011820731). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.