missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, 90+%, Thermo Scientific™

Product Code. p-7049970
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11358347

Brand: Thermo Scientific Alfa Aesar 032320.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 210°C to 215°C
Density 1.250 g/mL
Boiling Point 340°C to 342°C
Flash Point 121°C (249°F)
Quantity 500 g
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
Solubility Information Insoluble in water.
Formula Weight 178.23
Percent Purity ≥90%
Physical Form Crystalline
Chemical Name or Material Anthracene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.