Learn More
Anthrone, 95%, pure, Thermo Scientific Chemicals
Catalog Number p-4813328
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
90-44-8
C14H10O
194.23
MFCD00001187
RJGDLRCDCYRQOQ-UHFFFAOYSA-N
anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
7018
CHEBI:33835
10H-anthracen-9-one
O=C1C2=CC=CC=C2CC2=CC=CC=C12
90-44-8
C14H10O
194.23
MFCD00001187
RJGDLRCDCYRQOQ-UHFFFAOYSA-N
anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
7018
CHEBI:33835
10H-anthracen-9-one
O=C1C2=CC=CC=C2CC2=CC=CC=C12
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 152.0°C to 157.0°C
Boiling Point 721.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 07,473
Merck Index 15,682
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot naoh solution, conc. sulfuric acid, soluble in alcohol, benzene, aceton, soluble in most organic solvents without, fluorescence
Formula Weight 194.23
Percent Purity 95%
Grade Pure
Chemical Name or Material Anthrone
Show More Show Less
compliance-icons
Product Identifier
  • Anthrone
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.