Learn More
AAM, Mouse, Clone: 2H2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AAMP.
Brand: Abnova H00000014-M02.100ug
Description
The gene product is an immunoglobulin-type protein. It is found to be expressed strongly in endothelial cells, cytotrophoblasts, and poorly differentiated colon adenocarcinoma cells found in lymphatics. The protein contains a heparin-binding domain and mediates heparin-sensitive cell adhesion. [provided by RefSeq
Sequence: GEDDKAFVWRLSDGELLFECAGHKDSVTCAGFSHDSTLVATGDMSGLLKVWQVDTKEEVWSFEAGDLEWMEWHPRAPVLLAGTADGNTWMWKVPNGDCKTSpecifications
| AAM | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001087 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 14 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 2H2 | |
| Mouse monoclonal antibody raised against a partial recombinant AAMP. | |
| AAMP | |
| AAMP | |
| AAMP (NP_001078.2, 111 a.a. ∼ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.