Learn More
ABL2, Mouse, Clone: 6D5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ABL2.
Brand: Abnova H00000027-M03.100ug
Description
This gene encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. This gene is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq
Sequence: KKTLGLRAGKPTASDDTSKPFPRSNSTSSMSSGLPEQDRMAMTLPRNCQRSKLQLERTVSTSSQPEENVDRANDMLPKKSEESAAPSRERPKAKLLPRGASpecifications
| ABL2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC065912 | |
| ABL2 | |
| ABL2 (AAH65912, 743 a.a. ∼ 842 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, In situ PLA, KnockDown, Western Blot | |
| 6D5 | |
| Mouse monoclonal antibody raised against a partial recombinant ABL2. | |
| ABL2 | |
| ABLL/ARG/FLJ22224/FLJ31718/FLJ41441 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 27 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.