Learn More
acyl-Coenzyme A dehydrogenase, very long chain, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ACADVL.
Brand: Abnova H00000037-A01.50uL
Description
The protein encoded by this gene is targeted to the inner mitochondrial membrane where it catalyzes the first step of the mitochondrial fatty acid beta-oxidation pathway. This acyl-Coenzyme A dehydrogenase is specific to long-chain and very-long-chain fatty acids. A deficiency in this gene product reduces myocardial fatty acid beta-oxidation and is associated with cardiomyopathy. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: AAALAGTMRGIIAKAVDHATNRTQFGEKIHNFGLIQEKLARMVMLQYVTESMAYMVSANMDQGATDFQIEAAISKIFGSEAAWKVTDECISpecifications
| acyl-Coenzyme A dehydrogenase, very long chain | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ACADVL. | |
| ACADVL | |
| ACAD6/LCACD/VLCAD | |
| Mouse | |
| 50 μL | |
| Yes | |
| 37 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_000018 | |
| ACADVL | |
| ACADVL (NP_000009, 345 a.a. ∼ 434 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.