missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Actin, Monoclonal (Clone C4)

Product Code. 11373069 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373069 0.2 mL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11373069 Supplier MP Biomedicals Supplier No. 08691002
  • ONLINE EXCLUSIVE PRICE
    € 562.00
    € 534.45 / 200µL
    Save € 27.55
    4% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Anti-Actin, Monoclonal (Clone C4), is a mouse IgG1 (gamma 1 heavy/kappa light chain) antibody.

  • Chicken gizzard actin has been used as the immunogen
  • Immunoblots demonstrate specificity directed towards all actins ranging from yeast, to flies, to mouse, to man
  • Actins are: skeletal alpha-actin, cardiac alpha-actin, smooth muscle gamma actin, cytoplasmic beta-actin, cytoplasmic gamma actin as well as Dictyostelium discoideum and Physarum olycephalum actins
  • Epitope for the antibody lies between residues 18 to 40, a highly conserved region of the molecule
  • Can also be used for cells cultivated on slides, for frozen tissue sections and as a loading control for western blots

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen Actin
Classification Monoclonal
Conjugate Unconjugated
Host Species Mouse
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Product Type Antibody
Isotype IgG1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.