Learn More
adhesion regulating molecule 1, Mouse, Polyclonal Antibody, Abnova™
Description
The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | adhesion regulating molecule 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length recombinant ADRM1. |
| Formulation | 50% glycerol |
| Gene | ADRM1 |
| Gene Accession No. | BC017245 |
| Gene Alias | ARM1/GP110/MGC29536/Rpn13 |
| Gene Symbols | ADRM1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.