Learn More
Aiolos Mouse anti-Human, Brilliant Violet 421, Clone: S50-895, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565217
Description
The S50-895 monoclonal antibody specifically recognizes the transcription factor, Aiolos, which is encoded by IKZF3 , IKAROS family zinc finger 3. Aiolos belongs to the Ikaros family of zinc-finger proteins that includes Ikaros and Helios. Aiolos can form homodimers or heterodimers with other Ikaros family members. Multiple Aiolos isoforms have been described. Aiolos is expressed in committed lymphoid progenitors and mature types of T and B lymphocytes, but is not expressed by hematopoietic stem cells. Aiolos plays an important role in the control of B lymphocyte differentiation and proliferation and is specifically required for the generation of long-lived, high affinity plasma cells in the bone marrow. Aiolos is involved in regulating BCL2 expression and controlling apoptosis in T-cells in an IL2-dependent manner. Aberrant expression of IKZF3 has been implicated with leukemogenesis and certain autoimmune diseases.
Specifications
| Aiolos | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
| Human Aiolos Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| S50-895 | |
| AIOLOS; AIO; IKZF3; ZNFN1A3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.