Learn More
AKR7A2, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00008574-PW2.1Set
Product Code. 16182844
  • € 843.00 / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-AKR7A2 (300μg), Antibody pair for WB: rabbit polyclonal anti-AKR7A2 (50μL)
Specifications
AKR7A2
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
AFAR/AFAR1/AFB1-AR1/AKR7
Mouse, Rabbit
RUO
8574
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
AKR7A2
AKR7A2
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-AKR7A2 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-AKR7A2 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.