missing translation for 'onlineSavingsMsg'
Learn More
Learn More
aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human AKR7A2 protein.
Brand: Abnova H00008574-D01P.100ug
This item is not returnable.
View return policy
Description
Aldo-keto reductases, such as AKR7A2, are involved in the detoxification of aldehydes and ketones.[supplied by OMIM
Sequence: MSRPPPPRVASVLGTMEMGRRMDAPASAAAVRAFLERGHTELDTAFMYSDGQSETILGGLGLGLGGGDCRVKIATKANPWDGKSLKPDSVRSQLETSLKRLQCPQVDLFYLHAPDHGTPVEETLHACQRLHQEGKFVELGLSNYASWEVAEICTLCKSNGWILPTVYQGMYNATTRQVETELFPCLRHFGLRFYAYNPLAGGLLTGKYKYEDKDGKQPVGRFFGNSWAETYRNRFWKEHHFEAIALVEKALQAAYGASAPSVTSAALRWMYHHSQLQGAHGDAVILGMSSLEQLEQNLAATEEGPLEPAVVDAFNQAWHLVAHECPNYFRSpecifications
| aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human AKR7A2 protein. | |
| AKR7A2 | |
| AFAR/AFAR1/AFB1-AR1/AKR7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8574 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC010852.1 | |
| AKR7A2 | |
| AKR7A2 (AAH10852.1, 1 a.a. ∼ 330 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction