missing translation for 'onlineSavingsMsg'
Learn More

AMELX, Mouse, Clone: 3B5, Abnova™

Product Code. 16134614
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16134614 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16134614 Supplier Abnova Supplier No. H00000265M03A.200uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a partial recombinant AMELX.

This gene encodes a member of the amelogenin family of extracellular matrix proteins. Amelogenins are involved in biomineralization during tooth enamel development. Mutations in this gene cause X-linked amelogenesis imperfecta. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: PVIPQQPMMPVPGQHSMTPIQHHQPNLPPPAQQPYQPQPVQPQPHQPMQPQPPVHPMQPLPPQPPLPPMFPMQPLPPMLPDLTLEAWPSTDKTKREEVD

Specifications

Antigen AMELX
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant AMELX.
Formulation ascites with no preservative
Gene AMELX
Gene Accession No. NM_001142
Gene Alias AIH1/ALGN/AMG/AMGL/AMGX
Gene Symbols AMELX
Host Species Mouse
Immunogen AMELX (NP_001133, 93 a.a. ∼ 191 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 265
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.