Learn More
angiomotin, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant AMOT.
Brand: Abnova H00154796-A01.50uL
Description
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predominantly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: RKEPSKTEQLSCMRPAKSLMSISNAGSGLLSHSSTLTGSPIMEEKRDDKSWKGSLGILLGGDYRAEYVPSTPSPVPPSTPLLSAHSKTGSRDCSTQTERGSpecifications
| angiomotin | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant AMOT. | |
| AMOT | |
| KIAA1071 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 154796 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_133265 | |
| AMOT | |
| AMOT (NP_573572, 361 a.a. ∼ 460 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.