Learn More
Amyloid beta 40 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA316760
Additional Details : Weight : 0.01000kg
Description
Amyloid beta-protein (abeta) is associated with neuronal injury and death in Alzheimer′s disease. These proteins are the main components of amyloid plaques.Specifications
Amyloid beta 40 | |
Polyclonal | |
APP | |
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN2 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
P05067, P12023 | |
APP | |
Synthetic peptide corresponding to amino acid residues G L M V G G VV of human abeta 40. | |
RUO | |
11820, 351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |