Learn More
Androgen Receptor Mouse anti-Human, Unlabeled, Clone: G122-434, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554225
Description
The androgen receptor (AR) mediating androgen effects is a member of the steroid and thyroid hormone receptor gene superfamily encoding liganddependent nuclear transcription factors. They can be divided into three main domains, an N-terminal domain which modulates transcription efficiency, a central DNA domain which binds to a target gene hormone response element, and a C-terminal hormone binding domain. Androgens are necessary for normal male development and function. They exert their effects on target tissue through binding to the AR, followed by association of the AR complex with specific binding sites on DNA. This AR complex may induce or repress gene transcription. Mutations in the AR gene are associated with androgen insensitivity syndrome, a disorder that causes XY genotypic males to develop as phenotypic females because of their inability to respond to androgens. AR mutations have also been identified in human prostate cancer specimens and the LNCaP human prostate cancer cell line. Monoclonal and polyclonal antibodies to the AR have identified AR-positive cells in a variety of tissues including male and female sexual organs, kidney, liver, adrenal cortex, pituitary gland, skeletal, cardiac, and smooth muscle cells. The androgen receptor has been identified as a 100 kDa protein in SDS/PAGE. G122-434 recognizes an epitope localized within the N-terminal domain (between amino acids 33 and 485) of the human androgen receptor protein. The antibody does not recognize estrogen or progesterone receptors. Purified recombinant human androgen receptor protein was used as immunogen.
Specifications
| Androgen Receptor | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Purified human androgen receptor recombinant protein | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG2a |
| Western Blot | |
| G122-434 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.