missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ANGPTL3, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00027329-B01P.50ug
This item is not returnable.
View return policy
Specifications
| ANGPTL3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ANGPTL3 protein. | |
| ANGPTL3 | |
| ANGPT5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 27329 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014495 | |
| ANGPTL3 | |
| ANGPTL3 (NP_055310, 1 a.a. ∼ 460 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction