Learn More
Annexin VII Mouse anti-Human, Unlabeled, Clone: 5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610668
Description
Annexin VII, also known as synexin, is a member of the Annexin family characterized by Ca2+ dependent phospholipid binding. Annexins have a four-fold internal repeat of about 70 amino acids that contains the Ca2+-binding sites. They also have a regulatory NH2-terminal region of 30-40 amino acids. Annexin VII mRNA has been detected in many tissues and is most abundant in brain, heart, skeletal muscle, and lung. Annexin VII mRNA exhibits a tissue dependent polymorphism due to alternative splicing which results in two different isoforms of 47kDa and 51kDa. In vitro, Annexin VII aggregates chromaffin granules and enhances membrane fusion in a Ca2+ and GTP-dependent manner. Although the physiological role of Annexin VII is not completely understood, it is believed to influence and regulate Ca2+ dependent events such as secretion at the plasma membrane.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Annexin VII | |
| Monoclonal | |
| 250μg/mL | |
| Synexin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 5 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.