Learn More
annexin A10, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ANXA10 protein.
Brand: Abnova H00011199-D01P.100ug
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined. [provided by RefSeq
Sequence: MFCGDYVQGTIFPAPNFNPIMDAQMLGGALQGFDCDKDMLINILTQRCNAQRMMIAEAYQSMYGRDLIGDMREQLSDHFKDVMAGLMYPPPLYDAHELWHAMKGVGTDENCLIEILASRTNGEIFQMREAYCLQYSNNLQEDIYSETSGHFRDTLMNLVQGTREEGYTDPAMAAQDAMVLWEACQQKTGEHKTMLQMILCNKSYQQLRLVFQEFQNISGQDMVDAINECYDGYFQELLVAIVLCVRDKPAYFAYRLYSAIHDFGFHNKTVIRILIARSEIDLLTIRKRYKERYGKSLFHDIRNFASGHYKKALLAICAGDAEDYSpecifications
| annexin A10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human ANXA10 protein. | |
| ANXA10 | |
| ANX14 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 11199 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_007193.3 | |
| ANXA10 | |
| ANXA10 (NP_009124.2, 1 a.a. ∼ 324 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.