Learn More
APCS, Mouse, Clone: 4E8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant APCS.
Brand: Abnova H00000325-M07.100ug
Description
The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo. [provided by RefSeq
Sequence: WESSSGIAEFWINGTPLVKKGLRQGYFVEAQPKIVLGQEQDSYGGKFDRSQSFVGEIGDLYMWDSVLPPENILSAYQGTPLPANILDWQALNYEIRGYVIIKPLVWVSpecifications
| APCS | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001639 | |
| APCS | |
| APCS (NP_001630, 117 a.a. ∼ 223 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 4E8 | |
| Mouse monoclonal antibody raised against a partial recombinant APCS. | |
| APCS | |
| MGC88159/PTX2/SAP | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 325 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.