Learn More
SPEG complex locus, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human APEG1 protein.
Brand: Abnova H00010290-B01.50uL
Description
Expression of this gene is thought to serve as a marker for differentiated vascular smooth muscle cells which may have a role in regulating growth and differentiation of this cell type. The encoded protein is highly similar to the corresponding rat and mouse proteins. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of only one variant has been defined. [provided by RefSeq
Sequence: MKPSPSQNRRSSDTGSKAPPTFKVSLMDQSVREGQDVIMSIRVQGEPKPVVSWLRNRQPVRPDQRRFAEEAEGGLCRLRILAAERGDAGFYTCKAVNEYGARQCEARLEVRGESpecifications
| SPEG complex locus | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human APEG1 protein. | |
| SPEG | |
| APEG1/BPEG/KIAA1297/MGC12676/SPEGalpha/SPEGbeta | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC006346.2 | |
| SPEG | |
| APEG1 (AAH06346.1, 1 a.a. ∼ 113 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 10290 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.