Learn More
apolipoprotein A-II, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human APOA2 protein.
Brand: Abnova H00000336-D01P.100ug
Description
This gene encodes apolipoprotein (apo-) A-II, which is the second most abundant protein of the high density lipoprotein particles. The protein is found in plasma as a monomer, homodimer, or heterodimer with apolipoprotein D. Defects in this gene may result in apolipoprotein A-II deficiency or hypercholesterolemia. [provided by RefSeq
Sequence: MKLLAATVLLLTICSLEGALVRRQAKEPCVESLVSQYFQTVTDYGKDLMEKVKSPELQAEAKSYFEKSKEQLTPLIKKAGTELVNFLSYFVELGTQPATQSpecifications
| apolipoprotein A-II | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human APOA2 protein. | |
| APOA2 | |
| APOA2 | |
| APOA2 (NP_001634.1, 1 a.a. ∼ 100 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001643 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 336 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.