Learn More
ADP-ribosylation factor related protein 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ARFRP1.
Brand: Abnova H00010139-A01.50uL
Description
The protein encoded by this gene is a membrane-associated GTP-ase and localizes to the plasma membrane. It is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) genes. The gene is located in a gene cluster that includes the a gene (M68) that is overexpressed in some tumors. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: ANKQDVETCLSIPDIKTAFSDCTSKIGRRDCLTQACSALTGKGVREGIEWMVKCVVRNVHRPPRQRDITSpecifications
| ADP-ribosylation factor related protein 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ARFRP1. | |
| ARFRP1 | |
| ARL18/ARP/Arp1 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| ELISA | |
| Unconjugated | |
| 50% glycerol | |
| NM_003224 | |
| ARFRP1 | |
| ARFRP1 (NP_003215, 133 a.a. ∼ 201 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10139 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.