missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCL1 Rat anti-Mouse, Clone: 290622, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA523994
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.
Specifications
| ASCL1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q02067 | |
| ASCL1 | |
| E. coli-derived recombit mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu) | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 290622 | |
| Unconjugated | |
| Ascl1 | |
| Achaete-scute homolog 1, ASH-1, hASH1, Class A basic helix-loop-helix protein 46, bHLHa46, ASCL1, ASH1, BHLHA46, HASH1, Mash-1, Mash1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 17172 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction