missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG5 autophagy related 5 homolog (S. cerevisiae), Mouse, Clone: 4B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ATG5.
Brand: Abnova H00009474-M08.100ug
This item is not returnable.
View return policy
Description
Sequence: PAEENGFRYIPFRIYQTTTERPFIQKLFRPVAADGQLHTLGDLLKEVCPSAIDPEDGEKKNQVMIHGIEPMLETPLQWLSEHLSYPDNFLHISIIPQPTDSpecifications
| ATG5 autophagy related 5 homolog (S. cerevisiae) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC002699 | |
| ATG5 | |
| ATG5 (AAH02699, 176 a.a. ∼ 275 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Apoptosis | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4B2 | |
| Mouse monoclonal antibody raised against a partial recombinant ATG5. | |
| ATG5 | |
| APG5/APG5-LIKE/APG5L/ASP/hAPG5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 9474 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction