missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATOH1, Mouse, Clone: 2B7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ATOH1.
Brand: Abnova H00000474-M16.100ug
This item is not returnable.
View return policy
Description
This protein belongs to the basic helix-loop-helix (BHLH) family of transcription factors. It activates E-box dependent transcription along with E47. [provided by RefSeq
Sequence: MSRLLHAEEWAEVKELGDHHRQPQPHHLPQPPPPPQPPATLQAREHPVYPPELSLLDSTDPRAWLAPTLQGICTARAAQYLLHSPELGASEAAAPRDEVDGRGELVRRSSGGASSSKSPGPVKVREQLCKSpecifications
| ATOH1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005172 | |
| ATOH1 | |
| ATOH1 (NP_005163, 1 a.a. ∼ 130 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2B7 | |
| Mouse monoclonal antibody raised against a full length recombinant ATOH1. | |
| ATOH1 | |
| ATH1/HATH1/MATH-1/bHLHa14 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 474 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction