missing translation for 'onlineSavingsMsg'
Learn More
Learn More
atonal homolog 1 (Drosophila), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ATOH1 protein.
Brand: Abnova H00000474-B01P.50ug
This item is not returnable.
View return policy
Description
This protein belongs to the basic helix-loop-helix (BHLH) family of transcription factors. It activates E-box dependent transcription along with E47. [provided by RefSeq
Sequence: MSRLLHAEEWAEVKELGDHHRQPQPHHLPQPPPPPQPPATLQAREHPVYPPELSLLDSTDPRAWLAPTLQGICTARAAQYLLHSPELGASEAAAPRDEVDGRGELVRRSSGGASSSKSPGPVKVREQLCKLKGGVVVDELGCSRQRAPSSKQVNGVQKQRRLAANARERRRMHGLNHAFDQLRNVIPSFNNDKKLSKYETLQMAQIYINALSELLQTPSGGEQPPPPPASCKSDHHHLRTAASYEGGAGNATAAGAQQASGGSQRPTPPGSCRTRFSAPASAGGYSVQLDALHFSTFEDSALTAMMAQKNLSPSLPGSILQPVQEENSKTSPRSHRSDGEFSPHSHYSDSDEASSpecifications
| atonal homolog 1 (Drosophila) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ATOH1 protein. | |
| ATOH1 | |
| ATH1/HATH1/MATH-1/bHLHa14 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005172.1 | |
| ATOH1 | |
| ATOH1 (NP_005163.1, 1 a.a. ∼ 354 a.a) full-length human protein. | |
| 50 μg | |
| Transcription Regulation | |
| 474 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction