Learn More
ATPase, H+/K+ exchanging, beta polypeptide, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ATP4B protein.
Brand: Abnova H00000496-B02.50uL
Description
The protein encoded by this gene belongs to a family of P-type cation-transporting ATPases. The gastric H+, K+-ATPase is a heterodimer consisting of a high molecular weight catalytic alpha subunit and a smaller but heavily glycosylated beta subunit. This enzyme is a proton pump that catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. It is also responsible for gastric acid secretion. This gene encodes the beta subunit of the gastric H+, K+-ATPase. [provided by RefSeq]
Sequence: MAALQEKKTCGQRMEEFQRYCWNPDTGQMLGRTLSRWVWISLYYVAFYVVMTGLFALCLYVLMQTVDPYTPDYQDQLRSPGVTLRPDVYGEKGLEIVYNVSDNRTWADLTQTLHAFLAGYSPAAQEDSINCTSEQYFFQESFRAPNHTKFSCKFTADMLQNCSGLADPNFGFEEGKPCFIIKMNRIVKFLPSNGSAPRVDCAFLDQPRELGQPLQVKYYPPNGTFSLHYFPYYGKKAQPHYSNPLVAAKLLNIPRNAEVAIVCKVMAEHVTFNNPHDPYEGKVEFKLKIEKSpecifications
| ATPase, H+/K+ exchanging, beta polypeptide | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ATP4B protein. | |
| ATP4B | |
| ATP6B | |
| Mouse | |
| 50 μL | |
| Yes | |
| 496 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_000705.2 | |
| ATP4B | |
| ATP4B (NP_000696.1, 1 a.a. ∼ 291 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.