Learn More
ATPase, H+ transporting, lysosomal accessory protein 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ATP6AP1 protein.
Brand: Abnova H00000537-D01P.100ug
Description
This gene encodes a component of a multisubunit enzyme (1mDa MW) that mediates acidification of eukaryotic intracellular organelles. Vacuolar ATPase (V-ATPase) is comprised of a cytosolic V1 (site of the ATP catalytic site) and a transmembrane V0 domain. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, and receptor-mediated endocytosis. The encoded protein of this gene is approximately 45kDa and may assist in the V-ATPase-mediated acidification of neuroendocrine secretory granules. [provided by RefSeq
Sequence: MMAAMATARVRMGPRCAQALWRMPWLPVFLSLAAAAAAAAAEQQVPLVLWSSDRDLWAPAADTHEGHITSDLQLSTYLDPALELGPRNVLLFLQDKLSIEDFTAYGGVFGNKQDSAFSNLENALDLAPSSLVLPAVDWYAVSTLTTYLQEKLGASPLHVDLATLRELKLNASLPALLLIRLPYTASSGLMAPREVLTGNDEVIGQVLSTLKSEDVPYTAALTAVRPSRVARDVAVVAGGLGRQLLQKQPVSPVIHPPVSYNDTAPRILFWAQNFSVAYKDQWEDLTPLTFGVQELNLTGSFWNDSFARLSLTYERLFGTTVTFKFILANRLYPVSARHWFTMERLEVHSNGSVAYFNASQVTGPSIYSFHCEYVSSLSKKGSLLVARTQPSPWQMMLQDFQIQAFNVMGEQFSYASDCASFFSPGIWMGLLTSLFMLFIFTYGLHMILSLKTMDRFDDHKGPTISLTQIVSpecifications
| ATPase, H+ transporting, lysosomal accessory protein 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human ATP6AP1 protein. | |
| ATP6AP1 | |
| 16A/ATP6IP1/ATP6S1/Ac45/CF2/MGC129781/VATPS1/XAP-3/XAP3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 537 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001183.4 | |
| ATP6AP1 | |
| ATP6AP1 (NP_001174.2, 1 a.a. ∼ 470 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.