Learn More
B2M, Mouse, Clone: 3B12, Abnova™
Mouse monoclonal antibody raised against a full-length native B2M.
Brand: Abnova H00000567-M03.100ug
Description
This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia
Specifications
| B2M | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| B2M | |
| B2M | |
| Native purified human B2M. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 3B12 | |
| Mouse monoclonal antibody raised against a full length native B2M. | |
| PBS with no preservative; pH 7.4 | |
| P61769 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 567 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.