Learn More
BAIAP2, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00010458-PW2.1Set
Product Code. 16153134
  • € 843.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-BAIAP2 (300μL), Antibody pair for WB: mouse purified polyclonal anti-BAIAP2 (50μg)
Specifications
BAIAP2
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
BAP2/IRSP53
Mouse, Rabbit
RUO
10458
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
BAIAP2
BAIAP2
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-BAIAP2 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-BAIAP2 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.